Anti-SIGLEC9 Mouse Monoclonal Antibody (Unconjugated) [clone: ]

供应商: BioLegend
BLEG351502EA 0 CNY
BLEG351502
Anti-SIGLEC9 Mouse Monoclonal Antibody (Unconjugated) [clone: ]
抗体
Type: Primary
Antigen: SIGLEC9
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR