Anti-CD170 Mouse Monoclonal Antibody (Unconjugated) [clone: 1A5]

供应商: BioLegend
BLEG352002EA 0 CNY
BLEG352002
Anti-CD170 Mouse Monoclonal Antibody (Unconjugated) [clone: 1A5]
抗体
Type: Primary
Antigen: CD170
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR