Anti-CD45.2 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: ]

供应商: BioLegend
BLEG109824EA 2775 CNY
BLEG109823 BLEG109824
Anti-CD45.2 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: ]
抗体
Type: Primary
Antigen: CD45.2
Clonality: Monoclonal
Clone: 0
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR