Anti-PDCD1 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ]

供应商: BioLegend
BLEG109110EA 0 CNY
BLEG109109 BLEG109110
Anti-PDCD1 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: ]
抗体
Type: Primary
Antigen: PDCD1
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR