Anti-CD27 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: ]

供应商: BioLegend
BLEG302822EA 0 CNY
BLEG302821 BLEG302822
Anti-CD27 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: ]
抗体
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone:
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Baboon, Cynomolgus, Rhesus, Squirrel Monkey
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR