Anti-CD45.2 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]
供应商: BioLegend
BLEG109806EA 0 CNY
BLEG109805 BLEG109806
Anti-CD45.2 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]
抗体
Type: Primary
Antigen: CD45.2
Clonality: Monoclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
现在订购