Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]

供应商: BioLegend
BLEG110708EA 0 CNY
BLEG110707 BLEG110708
Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]
抗体
Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: 0
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR