Anti-H-2Kb Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]

供应商: BioLegend
BLEG116508EA 0 CNY
BLEG116507 BLEG116508
Anti-H-2Kb Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: ]
抗体
Type: Primary
Antigen: H-2Kb
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR