Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (FITC (Fluorescein Isothiocyanate))

供应商: BioLegend
BLEG400108EA 0 CNY
BLEG400107 BLEG400108
Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (FITC (Fluorescein Isothiocyanate))
抗体
Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR