Rat IgG2a kappa Isotype Control [clone: RTK2758] (PE (Phycoerythrin)/Cy7®)

供应商: BioLegend
BLEG400521EA 0 CNY
BLEG400521 BLEG400522
Rat IgG2a kappa Isotype Control [clone: RTK2758] (PE (Phycoerythrin)/Cy7®)
抗体
Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone: RTK2758
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR