Rat IgG2b kappa Isotype Control [clone: RTK4530] (PE (Phycoerythrin)/Cy7®)

供应商: BioLegend
BLEG400618EA 0 CNY
BLEG400617 BLEG400618
Rat IgG2b kappa Isotype Control [clone: RTK4530] (PE (Phycoerythrin)/Cy7®)
抗体
Type: Control
Antigen: IgG2 β
Clonality: Monoclonal
Clone: RTK4530
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Rat
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR