Anti-TNFA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]

供应商: BioLegend
BLEG502912EA 0 CNY
BLEG502912 BLEG502913
Anti-TNFA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ]
抗体
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Baboon, Cynomolgus, Rhesus, Pig, Pigtailed Macaque, Sooty Mangabey
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR