Anti-CD11B (Integrin Alpha M Chain, Mac-1) Rat Monoclonal Antibody [clone: M1/70.15]

供应商: Serotec
SERTMCA74GEA 0 CNY
SERTMCA74G
Anti-CD11B (Integrin Alpha M Chain, Mac-1) Rat Monoclonal Antibody [clone: M1/70.15]
抗体
Type: Primary
Antigen: CD11B (Integrin Alpha M Chain, Mac-1)
Clonality: Monoclonal
Clone: M1/70.15
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Human, Rabbit
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR