Anti-Strep-Tag Mouse Monoclonal Antibody [clone: Strep-tag II] (HRP (Horseradish Peroxidase))

供应商: Serotec
SERTMCA2489PEA 0 CNY
SERTMCA2489P
Anti-Strep-Tag Mouse Monoclonal Antibody [clone: Strep-tag II] (HRP (Horseradish Peroxidase))
抗体
Type: Secondary
Antigen: Strep-Tag Classic
Clonality: Monoclonal
Clone: Strep-tag II
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Synthetic Peptide
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR