T-connectors, barbed
供应商: Cole Parmer
COLEWZ-06365-77EA 0 CNY
COLEWZ-06365-77
T-connectors, barbed
PP, translucent.
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low- pressure applications (generally less than 30 psi).
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low- pressure applications (generally less than 2068 mbar).