Dulbecco’s phosphate buffered saline (DPBS)
供应商: Lonza
LONZ17-513F
Dulbecco’s phosphate buffered saline (DPBS)
DPBS is used as a buffer system to maintain cell culture media in the physiological pH range of 7,0 to 7,6.
It serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.