Microplate washer, Immuno™ Wash 1575
供应商: Bio-Rad
Immuno™ Wash
BRAD170-7009EA 0 CNY
BRAD170-7009
Microplate washer, Immuno™ Wash 1575
微孔板垫圈
The ImmunoWash™ 1575 microplate washer is essential for performing accurate and reproducible immunoassays in a microplate format, such as ELISA. It is suitable for autoclave sterilisation and compatible with 96-well microplates that have flat-, U-, or V-bottom wells and strips.
  • Choose from a variety of preprogrammed wash sequences
  • Design, store, and reproducibly perform special wash routines
  • Maximise wash efficiency by adapting to various liquids and buffers, assay components and conditions
  • Exercise complete control of needle position (horizontal and vertical) and aspiration/dispensing rates
Dimensions: 345×423×200 mm
Weight: 10,3 kg