Shelf, roller
供应商: Metro International
METOCCROLWS-24EA 0 CNY
METOCCROLWS-24
Shelf, roller
Wire pullout shelves with roller pulls out halfway easily even at the bottom level where accessibility is vital. Each shelf includes two attachment brackets.
Weight capacity: 68 kg.