棉垫
供应商: VWR
VWR®
VWRU21902-985_UEA 0 CNY
VWRU21902-985_U
棉垫
拭子 清洗用拭子
经济、多用途的100%纯棉垫。垫子柔软,纯净,吸水性高,不会划伤最脆弱的涂层或表面。不会污染待培液或化学品。
  • 没有粘合剂或化学物质
  • 强度好,不管是干燥还是浸湿状态
Suitable for use in the printing industry.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904