VWR®, carboy,广口,矩形,带旋塞
供应商: VWR
VWR®
VWRI216-1732EA 1251.19 CNY
VWRI216-1732 VWRI216-1735 VWRI216-1700 VWRI216-1719
VWR®, carboy,广口,矩形,带旋塞
carboy水瓶
HDPE or PP translucent, with purple screw caps.
Use for storing and dispensing solutions and media. Rectangular carboys feature a heavy-duty stainless steel handle for easy carrying and pouring. Carboys are leakproof on standard test conditions. Graduated to identify the volume dispensed. Stopcocks are tied to the handle and require hand assembly for use. No tools are necessary.
  • 优异的耐化学性
  • 工作温度0-135℃
  • 可高温高压灭菌
PP carboys
Ideal for sterile water storage, polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Excellent chemical resistance
Working temperature: 0 to 135 °C
Autoclavable
Note: Polypropylene carboys are autoclavable. HDPE carboys are not autoclavable. Always remove stopcock and cap from the carboy before autoclaving.
高密度聚乙烯材质的一次性carboy,非常适合于存储和转移无菌液体以及制药/生物技术试剂。
Certifications: Resin meets US FDA 21 CFR, USP Class VI
  • 适合冷藏(低至-80℃),工作温度+120℃至-80℃
  • 即使在零下温度也能保持高冲击强度
  • 耐化学性极好,广泛用于长期存储
  • 广泛用于食品和乳品行业
注意:聚丙烯carboy可高温高压灭菌。HDPEcarboy不可进行高温高压灭菌。高温高压灭菌前,务必取下carboy的插塞和帽盖。