Dissolved oxygen sensor, VisiFerm DO Arc 225
供应商: Hamilton
HAMI242164EA 0 CNY
HAMI242164
Dissolved oxygen sensor, VisiFerm DO Arc 225
The VisiFerm DO Arc is the first optical oxygen sensor with integrated opto-electronics, having the full functionality of a measuring device with self-diagnostics.
  • Measurement principle: Oxygen dependent luminescence quenching
  • Steam sterilisable, autoclavable and CIP compatible
  • Requires less maintenance than a classical oxygen sensor as it does not have a mechanically sensitive membrane or a corrosive electrolyte