HPLC columns, Avantor® ACE® SuperC18™, microbore, 2 µm

供应商: Avantor®
Avantor® ACE® SuperC18
ACETACE-1011-0301EA 5940 CNY
ACETACE-1011-0301 ACETACE-101105005S ACETACE-1011-0501S ACETACE-1011-0301S ACETACE-1011-1001 ACETACE-101112005S ACETACE-101110005S ACETACE-1011-12005 ACETACE-1011-1501S ACETACE-1011-05005 ACETACE-1011-1501 ACETACE-1011-1201 ACETACE-1011-7501 ACETACE-1011-03005 ACETACE-1011-0501 ACETACE-1011-10005 ACETACE-101115005S ACETACE-101103005S ACETACE-1011-1001S ACETACE-1011-7501S ACETACE-1011-15005 ACETACE-101175005S ACETACE-1011-1201S ACETACE-1011-75005
HPLC columns, Avantor® ACE® SuperC18™, microbore, 2 µm
色谱柱
Avantor® ACE® SuperC18™ columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.

  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1,7 to 10 µm) and complementary phases

Maximum recommended pH: 1,5 to 11,5

Max. pressure: 275 bar
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR