HPLC/UHPLC columns, Avantor® ACE® Excel® CN, analytical, 5 µm
供应商: Avantor®
Avantor® ACE® Excel®
ACETEXL-124-2546UEA 5000 CNY
ACETEXL-124-2546U ACETEXL-124-1546U ACETEXL-124-1003U
HPLC/UHPLC columns, Avantor® ACE® Excel® CN, analytical, 5 µm
色谱柱
These Avantor® ACE® CN columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Can be used under both reversed phase and normal phase conditions
  • Capillary to preparative dimensions
  • Wide range of particle sizes (2, 3 and 5 µm)
  • Useful for the separation of polar analytes and compounds with double and triple bonds
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Where Avantor® ACE® Excel® CN columns are no longer available, please see the Avantor® ACE® CN product page for additional dimensions.