HPLC columns, Avantor® ACE® C18-PFP, 5 µm
供应商: Avantor®
ACE®
ACETACE-121003005SEA 10890 CNY
ACETACE-121003005S ACETACE-1210-05005 ACETACE-1210-0301S ACETACE-1210-0501S ACETACE-121010005S ACETACE-1210-1501 ACETACE-1210-1201 ACETACE-121015005S ACETACE-1210-25005 ACETACE-1210-1001 ACETACE-1210-7501S ACETACE-121075005S ACETACE-1210-7501 ACETACE-1210-1201S ACETACE-1210-3501 ACETACE-1210-75005 ACETACE-1210-35005 ACETACE-1210-1001S ACETACE-121005005S ACETACE-1210-12005 ACETACE-1210-10005 ACETACE-1210-2501S ACETACE-1210-0301 ACETACE-1210-2501 ACETACE-1210-03005 ACETACE-1210-30005 ACETACE-1210-1501S ACETACE-1210-15005 ACETACE-12100101GD ACETACE-121012005S ACETACE-1210-0501 ACETACE-121025005S
HPLC columns, Avantor® ACE® C18-PFP, 5 µm
色谱柱
Avantor® ACE® C18-PFP phase utilises a specially developed ligand combining a C18 chain with integral PFP functionality. This results in a phase that maintains the hydrophobic, stability and low bleed characteristics of leading C18 phases, yet provides the multiple retention mechanisms of a PFP phase.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 µm to 10 µm) and complementary phases
  • Also useful for analytes with π-bonding or electron or proton donor groups
  • Ultra-low bleed to ensure LC-MS compatibility
  • Method validation kits contain three different batches of the same phase - available on request
Due to its integral pentafluorophenyl functionality, Avantor® ACE® C18-PFP is also recommended for separations that involve halogenated aromatic compounds, regioisomers, and those analytes with differing shape constraints.
These columns are made from stainless steel.
订购须知 Method development and validation kits are available on request. Please contact chromsupport@avantorsciencs.com.