Microvolume Spectrophotometers, NanoDrop™ One, VWR-Peqlab (Thermo Scientific)
供应商: THERMO FISHER (NANODROP)
NanoDrop™
NANDND-ONE-WEA 0 CNY
NANDND-ONE-W
Microvolume Spectrophotometers, NanoDrop™ One, VWR-Peqlab (Thermo Scientific)
光谱仪 生物分光光度计
The NanoDrop One is designed to quantify and qualify DNA, RNA and protein samples.
  • Quick collection of data with auto-blank and auto-measure modes
  • Organisation of DNA quantification data
  • Ergonomic stand-alone design with integrated Android™ tablet
Identify contaminants, obtain corrected concentrations for DNA quantitation.
Accessories information: No implementation by VWR Service Team included !