VITEK® 2 compact identification cards
供应商: bioMérieux Industry
BMER21342EA 0 CNY
BMER21342 BMER21343 BMER21341 BMER21347 BMER21345
VITEK® 2 compact identification cards
微生物学测试系统 微生物检验
VITEK® 2 Compact is an automated phenotypic identification system for routine microbial identification that provides less hands on time and quick identification results.
  • 64-well cards with advanced colorimetric substrates provide high accuracy
  • Each card has a unique barcode identifier linking it to its sample
  • Cards are hermetically sealed keeping the sample safe inside