Irradiated swabs, QUANTISWAB®
供应商: bioMérieux Industry
BMER43801EA 0 CNY
BMER43801
Irradiated swabs, QUANTISWAB®
拭子 采样用拭子
This irradiated nylon flocked swab is made from micro-fibres of Nylon projected against an adhesive tip.
  • Great versatility with either direct inoculation immediately after swabbing, or use the included transport tube
  • 90% release capacity of the captured microorganisms for reliable and improved microorganism recovery from surfaces
  • Capable of performing quantitative environmental monitoring