Weighing bottles, glass, PYREX®
供应商: Corning
PYREX®
CORN1680-3060EA 0 CNY
CORN1680-4050 CORN1680-3060 CORN1680-5060 CORN1680-4080 CORN1682-6030 CORN1682-7033 CORN1682-5030 CORN1680-2550 CORN1680-2540 CORN1680-1580 CORN1680-1550
Weighing bottles, glass, PYREX®
称量瓶
Made of PYREX® borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
These tall or low form bottles have a standard taper joint style opening that accepts a closed bottom, hollow pennyhead style stopper.