Burettes, PTFE stopcock, serialised, PYREX®
供应商: Corning
PYREX®
CORN2105-50EA 0 CNY
CORN2105-50 CORN2105-100 CORN2105-25 CORN2105-10
Burettes, PTFE stopcock, serialised, PYREX®
滴定管
Borosilicate glass, class A.
  • Individually serialised
  • Coloured markings with fine sharp lines
  • Large easy to read numbers
  • Smooth micro-finish stopcock assures leak-resistant performance
Burette tips are carefully drawn from precision bore tubing to ensure proper drainage rates.
Designed as per ASTM E287 and calibrated in accordance with ASTM E542 and D-511 requirements. Traceable to NIST standards.
Delivery information: Delivery information: Burettes are supplied with 2 mm bore PTFE stopcock plug, a dust cover and certificate of identification and capacity.
Certifications: PYREX® Serialised/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Delivery information: Burettes are supplied with 2 mm bore PTFE stopcock plug, a dust cover and certificate of identification and capacity.