Watch glasses, borosilicate glass
供应商: Corning
Corning®
CORN9985-150EA 0 CNY
CORN9985-150 CORN9985-125_U CORN9985-65 CORN9985-75 CORN9985-125 CORN9985-90 CORN9985-100
Watch glasses, borosilicate glass
观察镜
PYREX® borosilicate glass.
  • High chemical durability
  • Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.