Stoppers, Standard Taper, HDPE, PYREX®
供应商: Corning Life Sciences
Corning®
CORN7624-32EA 0 CNY
CORN7624-22 CORN7624-13 CORN7624-19 CORN7624-32 CORN7624-16 CORN7624-27 CORN7624-38 CORN7624-8 CORN7624-9
Stoppers, Standard Taper, HDPE, PYREX®
塞子 聚乙烯/低密度聚乙烯/高密度聚乙烯塞
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
  • Blue and white with a closed bottom
  • Three rings on the plug provide an efficient seal
  • Improved design fully protects the neck from accidental tipping