VWR®, Biohazard Bags
供应商: VWR
VWR®
VWRU14220-016EA 303.35 CNY
VWRU14220-016 VWRU14220-004 VWRU14220-018 VWRU14220-006 VWRU14220-008 VWRU14220-010 VWRU14220-012 VWRU14220-000 VWRU14220-014 VWRU14220-024 VWRU14220-002
VWR®, Biohazard Bags
废物处理袋
These bags provide an efficient, safe solution for the messy and potentially dangerous process of disposing of biohazardous waste material.
  • PP, 38 µm thick
  • Perfect adaptability to standard laboratory disposal routines, including steam autoclaving
  • With biohazard warning and printed area
  • For use at 135 °C
Delivery information: Twist ties are included.