VWR®, Oil for Vacuum Pump
供应商: Avantor
VWR®
VWRU54996-061EA 234.42 CNY
VWRU54996-082 VWRU54996-061
VWR®, Oil for Vacuum Pump
润滑油
Superior oil based upon special crude oil stocks noted for their durability and fine lubricating qualities.
  • Durable
  • Fine lubricating qualities
  • Clean oil runs coolly in mechanical pumps
  • Improves system lifetime
The oil is further enhanced through precision distilling, resulting in a cleaner oil that runs cooler in most mechanical pumps. Cooler operation reduces thermal cracking for longer oil life. Good forepressures are prolonged, reducing backstreaming and there are often noticeable improvements in pumping cycle times.