Crimp/snap top vials
供应商: VWR Collection
VWR®
VWRU82028-416EA 2576.25 CNY
VWRU66009-872 VWRU82028-416
Crimp/snap top vials
小瓶 色谱瓶
These borosilicate glass vials feature a versatile neck that allows closure by either snap-cap seals or aluminium crimp seals. They have 40% wider openings than standard vials to reduce risk of needle damage. Compatible with Agilent® 7673 and 1100 series autosamplers.
Delivery information: Convenience kits include the vials and unassembled snap caps. Supplied in dust-free storage trays.