First aid kit
供应商: VWR Collection
VWRU56613-218EA 342 CNY
VWRU56613-218 VWRU56613-217
First aid kit
Portable first aid kit for laboratory emergencies.
  • Sterile items are individually wrapped
  • Contents are protected from moisture, dust, and other contaminants by gasketed seal
  • First aid guide is included
  • Packaged in a sturdy metal case
  • Screw holes are provided in back panel for wall mounting in visible locations
Content:
1×cold pack
1×adhesive bandage without adhesive pad
10×adhesive tesla pads (5×7,6 cm)
2×sterile bandage rolls (5 cm × 3,7 m)
1×triangular compress bandage
10×gauze pads (7,6×7,6 cm)
1×ABD combination pad (12,7×22,9 cm)
3×ammonia inhalants
2×eye pads
1×adhesive tape roll (1,3 cm × 4,6 m)
6×PVP iodine wipes
5×¹/₈ g Bu/rn Jel® pain relief pouches
One pair of angled scissors
One pair of pointed plastic forceps
One pair of latex gloves
One first aid guide