VWR®, Absorbent mat pads and rolls
供应商: VWR
VWR®
VWRU82028-028EA 2443.61 CNY
VWRU82028-028 VWRU82028-018
VWR®, Absorbent mat pads and rolls
泄露控制装置
Absorbent pads and rolls are ideal for spill clean-up; fast-wicking dimpled design absorbs liquids quickly to leave area clean and dry for improved safety. Efficient high-absorbency materials reduce waste of absorbents.
  • Soak up leaks and spills fast, keeping floors at your workplace clean, dry and safe
  • High-capacity absorbents stay on the job longer and are perfect for stocking spill kits
  • Can be incinerated after use to reduce waste
Absorbent pads and rolls are ideal for spill clean-up; fast-wicking dimpled design absorbs liquids quickly to leave area clean and dry for improved safety. Efficient high-absorbency materials reduce waste of absorbents. These general purpose pads and rolls absorb water-based liquids, petroleum products and solvents for everyday maintenance and cleanup.
  • Soak up leaks and spills fast, keeping floors at your workplace clean, dry and safe
  • High-capacity absorbents stay on the job longer and are perfect for stocking spill kits
  • Can be incinerated after use to reduce waste
These general purpose pads and rolls absorb water-based liquids, petroleum products and solvents for everyday maintenance and cleanup.