Tubing, PVC, clear
供应商: VWR Collection
VWR®
VWRU60985-528EA 598 CNY
VWRU60985-530 VWRU60985-528
Tubing, PVC, clear
胶管 聚氯乙烯管材
PVC, clear.
  • Resists a wide range of chemicals
  • Made from the highest grade resins and special plasticisers (no fillers or extenders are added)
  • Tensile strength of 113,7 bar
Ideal for use in medical research, clinical, pharmaceutical, food and beverage, media manufacture and transfer, and vapour transfer applications. Can also be used with peristaltic pumps, CO₂ incubators, and condenser coolant lines. This autoclavable, lab-grade clear plastic tubing is dimensionally stable and highly resistant to hardening and discoloration.
Shore hardness: 55° (A)
Certifications: Complies with FDA, USP Class VI and USDA standards.