Test tube racks, wire, epoxy-coated, VWR®
供应商: VWR Collection
VWR®
VWRU60916-704EA 478 CNY
VWRU60916-704 VWRU60916-884
Test tube racks, wire, epoxy-coated, VWR®
架子 Tube Racks
Colour-coded rectangular racks features square holes, three tiers for tube support and a wire-lined bottom. Orange racks can be used for biohazard warning and yellow for radioactive warning.
  • Chemical and corrosion-resistant
  • Withstand a temperature range of –223 to +121 °C