Test tube racks, wire, epoxy-coated
供应商: VWR Collection
VWR®
VWRU60916-704EA 478 CNY
VWRU60916-884 VWRU60916-704
Test tube racks, wire, epoxy-coated
架子 试验管架
Colour-coded rectangular racks features square holes, three tiers for tube support and a wire-lined bottom. Orange racks can be used for biohazard warning and yellow for radioactive warning.
  • Chemical and corrosion-resistant
  • Withstand a temperature range of –223 to +121 °C