VWR®, Beaker Tongs, Carbon Steel
供应商: VWR
VWRU82027-360EA 661.32 CNY
VWRU82027-360
VWR®, Beaker Tongs, Carbon Steel
钳子
Carbon steel, cadmium plated, with plastic coated tips.
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 ml capacities.