VWR®, Measuring Cylinders, Tall Form, Class B, Borosilicate Glass
供应商: VWR
VWR®
VWRU89000-254EA 216.45 CNY
VWRU89000-254 VWRU89000-252 VWRU89000-262 VWRU89000-260 VWRU89000-248 VWRU89000-256
VWR®, Measuring Cylinders, Tall Form, Class B, Borosilicate Glass
量筒 Measuring Cylinders
Borosilicate glass, tall form, class B.
  • Calibrated to contain at 20 °C ( TC)
  • Graduations printed in durable white enamel
  • Hexagonal base provides stability
Glass conforming to ASTM E1272, Style 1, Class B requirements.
Delivery information: Supplied with a LDPE SAFE-GARD® bumper on sizes 25 ml and larger.