VWR® Talon®, Burette clamp
供应商: VWR
VWR® Talon®
VWRU17683-258EA 888.38 CNY
VWRU17683-258
VWR® Talon®, Burette clamp
夹具 通用夹具
Zinc. This double burette clamp holds any size burette from micro to 100 ml capacity. Simply compress the scissor-like mechanism, insert burette, and gently release to grip.
  • Numbers and graduation on burette remain easy to read
  • For height adjustments, recompress mechanism and slide buret up or down and gently release
This double burette clamp holds any size burette from micro to 100 ml capacity. Simply compress the scissor-like mechanism, insert burette, and gently release to grip.
  • Numbers and graduation on burette remain easy to read
  • For height adjustments, recompress mechanism and slide buret up or down and gently release