Filtering flasks, graduated, with glass hose connection
供应商: VWR Collection
VWR®
VWRU89000-386EA 4275 CNY
VWRU89000-384 VWRU89000-376 VWRU89000-386 VWRU89000-382 VWRU89000-378
Filtering flasks, graduated, with glass hose connection
烧瓶 过滤瓶
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These flasks are designed to take 1 atmosphere of vacuum or 29" of mercury. All sizes have side hose connection designed to accept ⁵/₁₆" int.Ø. flexible tubing. Sizes 1000 ml and larger accept 9,5 mm int.Ø tubing.
Certifications: Meet ASTM specification E1406, Type II, Class I requirements.