Disposal box for pipette tips
供应商: VWR Collection
VWR®
VWRU82010-754EA 2173.81 CNY
VWRU82010-754
Disposal box for pipette tips
箱子 β/γ辐射箱
Acrylic, clear
  • Eliminates any need to handle pipette tips used with hazardous fluids
  • The 10 mm thick acrylic provides protection from beta radiation
  • Large, hinged lid allows easy removal of contents
This box features a flip-top port cover for convenient one-handed disposal of used pipette tips.