VWR®, Powder Funnels
供应商: VWR, part of Avantor
VWR®
VWRU89000-474EA 1784.25 CNY
VWRU89000-474 VWRU89000-470 VWRU89000-466
VWR®, Powder Funnels
漏斗 粉末漏斗
Borosilicate glass 3.3.
  • Constructed with heavy uniform wall
  • Strong fire-polished rim and stem
  • High resistance to chemical attack, mechanical and thermal shock
Certifications: Conform to ASTM specification E1095, Type IV requirements.