Jars, glass, clear, wide mouth
供应商: VWR Collection
VWR®
VWRU89043-268EA 605 CNY
VWRU89043-268 VWRU89043-382 VWRU89043-266 VWRU89043-388 VWRU89043-380 VWRU89043-272 VWRU89043-296 VWRU89043-370 VWRU89043-290 VWRU89043-300 VWRU89043-294 VWRU89043-384 VWRU89043-298 VWRU89043-270
Jars, glass, clear, wide mouth
通用罐
Soda-lime glass, clear.
Designed for storage without shoulders to maximize capacity for contents.
  • Wide mouth design for efficient addition and removal of contents
  • Clear glass allows for easy viewing of contents
Designed for storage without shoulders to maximise capacity for contents.
  • Wide mouth design for efficient addition and removal of contents
  • Clear glass allows for easy viewing of contents
Conforms to USP Type III and ASTM E438, Type II requirements.
Certifications: Conforms to USP Type III and ASTM E438, Type II requirements.
Delivery information: Convenience packs consist of corrugated cases with dividers separating glassware in smaller quantities (12 or 24 per pack). Bulk packs consist of shrink-wrapped glassware bundles (12 to 24 per wrap) packaged in corrugated cases. Caps are included in separate bags within the case. Jars sold without caps are packaged in bulk packs. Jars with capacity of 1000 ml are packed in corrugated cartons with divider cells.