Bulb pipettes, class A
VWRU89045-060EA 960.5 CNY
VWRU89045-060 VWRU89045-070 VWRU89045-062 VWRU89045-084 VWRU89045-072 VWRU89045-064 VWRU89045-086 VWRU89045-074 VWRU89045-066 VWRU89045-076 VWRU89045-054 VWRU89045-080 VWRU89045-068 VWRU89045-056 VWRU89045-078 VWRU89045-058
Bulb pipettes, class A
Reusable
  • Calibrated to deliver
  • Sizes 1 ml and larger are marked with large numerals on the bulb, indicating capacity
  • Color-coded per ASTM specification E1273 for ease in sorting and selecting the correct pipet
  • Graduations of permanent brown stain
Certifications: Designed from ASTM specification E969.