Chemistry kit, PYREX®
供应商: Corning
PYREX® Corning®
CORN6949EA 0 CNY
CORN6949N-IO CORN8950-24 CORN2490-24 CORN9601-24 CORN6949N-BO CORN6949 CORN2401-24 CORN9021-24
Chemistry kit, PYREX®
微量化学试剂
Chemistry kit is designed for most common organic chemistry experiments.
It comprises 15 components with 24/40 standard tapered joints including flasks, adapters, condensers, condenser column, funnel, stopper and reaction vessel. The components are held in foam within a polyethylene case.