Safety bottle jacket
供应商: Justrite
JUSU12009EA 0 CNY
JUSU12009
Safety bottle jacket
搬运篮 瓶载体/容器
PE, translucent
  • Translucent colour aids in detecting liquid level
  • Jacket acts as a shock absorber, minimising breakage and spills due to bumps or falls
Bottles fit into the bottom half of the jacket and lid twists on securely for safe pouring.