Spoon spatula
供应商: VWR, part of Avantor
VWR®
VWRU82027-532EA 88.56 CNY
VWRU82027-532
Spoon spatula
样品勺
Stainless steel. This double spatula is ideal for use in the transfer, collection, and handling of chemical or biological samples.
  • Rustproof stainless steel
  • Flat spatula at one end and a spoon at the other
  • Substances will not stick to spatula surfaces