Bunsen burner
供应商: VWR Collection
VWRU89038-530EA 432 CNY
VWRU89038-532 VWRU89038-530
Bunsen burner
加热器 煤气灯
Bunsen burner with nickel plated zinc alloy die-cast base.
  • Designed with a flame stabiliser to provide a steady flame for general laboratory needs
  • With aluminium mixing tubes and serrated gas inlet and air regulator
  • Optional wing top produces a broad flat flame for working glass and general lab use
Height: 156 mm
订购须知 Wing top must be ordered separately.